Learn More
AdipoGen Cyflufenamid
SDP
Supplier:  AdipoGen CDXC0230M500
Encompass_Preferred
Chemical. CAS 180409-60-3. Formula C20H17F5N2O2. MW 412.36. Synthetic Amidoxime fungicide used in agroculture. Active against powdery mildew of various crops in pre and post infection treatment. Has a long lasting translaminar and vapor phase activity. The mode of action is not know and it is considered different from that of other existing fungicides. It has favorable toxicological, ecotoxicological and environmental profiles. Compound can be used as analytical reference material.
Catalog No. 50-256-1423