Learn More
Medchemexpress LLC β-Cyfluthrin | 1820573-27-0 | 99.9% | 100 MG
SDP
Catalog No. 5000410610
Encompass_Preferred
β-Cyfluthrin is a type II synthetic pyrethroid and an active ingredient in many insecticide products used in agriculture. It acts as a neurotoxicant, influencing calcium concentration in nervous tissue by inhibiting Ca2+ ATPase, which is involved in calcium transport.
  • High purity
  • Targets Ca2+ ATPase
  • Reduces cell viability in primary rat hepatocytes at 80 μM over 48 hours, impacting intracellular malondialdehyde (MDA) and nitrate/nitrite levels, and decreasing superoxide dismutase (SOD) activity
  • In mouse models, administration of 20-40 mg/kg (po, once daily for 40 days) inhibits locomotor activity, impairs learning ability, and causes anxiety-like behaviors and growth retardation
Catalog No. 50-004-10610 Supplier Medchemexpress LLC Supplier No. HYB1837A100MG
β-Cyfluthrin is a type II synthetic pyrethroid and an active ingredient in many insecticide products used in agriculture. It acts as a neurotoxicant, influencing calcium concentration in nervous tissue by inhibiting Ca2+ ATPase, which is involved in calcium transport.
  • High purity
  • Targets Ca2+ ATPase
  • Reduces cell viability in primary rat hepatocytes at 80 μM over 48 hours, impacting intracellular malondialdehyde (MDA) and nitrate/nitrite levels, and decreasing superoxide dismutase (SOD) activity
  • In mouse models, administration of 20-40 mg/kg (po, once daily for 40 days) inhibits locomotor activity, impairs learning ability, and causes anxiety-like behaviors and growth retardation