Learn More
Cayman Chemical CyfluthrIn-d6 1.2mL
SDP
Catalog No. 502919184
Encompass_Preferred
An internal standard for the quantification of cyfluthrin by GC- or LC-MS
Catalog No. 50-291-9184 Supplier Cayman Chemical Supplier No. 396001.2 ML
Add to Cart
Add to Cart
An internal standard for the quantification of cyfluthrin by GC- or LC-MS