Learn More
Medchemexpress LLC CYGB Rat His-Myc 10ug
SDP
Catalog No. 5000183239
Encompass_Preferred
Recombinant rat cytoglobin (CYGB) fused to His and Myc affinity tags, supplied as a purified protein for research applications. This preparation is intended for biochemical assays, binding studies, and antibody validation; it is provided in microgram-scale quantities and shipped on cold packs to preserve integrity. Research use only.
  • Purified recombinant protein suitable for biochemical assays and antibody validation.
  • N-terminal His tag and C-terminal Myc tag for purification and detection.
  • Approximate molecular weight 31 kDa enabling expected migration on SDS-PAGE.
  • Purity around 90% as reported by the supplier.
  • Supplied in microgram-scale amount and shipped on cold packs.
  • For research use only.
Catalog No. 50-001-83239 Supplier Medchemexpress LLC Supplier No. HYP7216210UG
Recombinant rat cytoglobin (CYGB) fused to His and Myc affinity tags, supplied as a purified protein for research applications. This preparation is intended for biochemical assays, binding studies, and antibody validation; it is provided in microgram-scale quantities and shipped on cold packs to preserve integrity. Research use only.
  • Purified recombinant protein suitable for biochemical assays and antibody validation.
  • N-terminal His tag and C-terminal Myc tag for purification and detection.
  • Approximate molecular weight 31 kDa enabling expected migration on SDS-PAGE.
  • Purity around 90% as reported by the supplier.
  • Supplied in microgram-scale amount and shipped on cold packs.
  • For research use only.