Learn More
Medchemexpress LLC Cyhalofop-butyl | 122008-85-9 | 98.7% | 10 MM * 1 ML
SDP
Catalog No. 5000408073
Encompass_Preferred
Cyhalofop-butyl is a post-emergence herbicide that inhibits acetyl-coenzyme A carboxylase (ACCase) biosynthesis. It has a molecular weight of 357.38 and a chemical formula of C20H20FNO4.
  • Post-emergence herbicide
  • Inhibits ACCase biosynthesis
  • White to off-white solid appearance
  • Soluble in DMSO (250 mg/mL)
  • Store solid at 4°C, protected from light
  • Store solutions at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-08073 Supplier Medchemexpress LLC Supplier No. HYB086110MM/1ML
Cyhalofop-butyl is a post-emergence herbicide that inhibits acetyl-coenzyme A carboxylase (ACCase) biosynthesis. It has a molecular weight of 357.38 and a chemical formula of C20H20FNO4.
  • Post-emergence herbicide
  • Inhibits ACCase biosynthesis
  • White to off-white solid appearance
  • Soluble in DMSO (250 mg/mL)
  • Store solid at 4°C, protected from light
  • Store solutions at -80°C for 6 months or -20°C for 1 month, protected from light