Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyhalofop-butyl | 122008-85-9 | 98.7% | 50 MG
SDP

Catalog No. 5000409414
Encompass_Preferred

Cyhalofop-butyl is a post-emergence herbicide that inhibits acetyl-coenzyme A carboxylase (ACCase) biosynthesis.

  • Purity of 98.7%
  • Appears as a white to off-white solid
  • Molecular weight of 357.38
  • Chemical formula C20H20FNO4
  • Ships at room temperature in continental US
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for up to 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 250 mg/mL

Catalog No. 50-004-09414 Supplier Medchemexpress LLC Supplier No. HYB086150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyhalofop-butyl is a post-emergence herbicide that inhibits acetyl-coenzyme A carboxylase (ACCase) biosynthesis.

  • Purity of 98.7%
  • Appears as a white to off-white solid
  • Molecular weight of 357.38
  • Chemical formula C20H20FNO4
  • Ships at room temperature in continental US
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for up to 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 250 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.