Learn More
Medchemexpress LLC λ-Cyhalothrin | 91465-08-6 | 99.9% | 250 MG
SDP
Catalog No. 5000405598
Encompass_Preferred
λ-Cyhalothrin is a highly efficient, broad-spectrum type II synthetic pyrethroid insecticide containing an α-cyano group. It is utilized to manage a wide array of pests across various applications. This neurotoxin targets sodium channels within the membranes of neurons in the central nervous system.
  • Highly efficient, broad-spectrum type II synthetic pyrethroid insecticide.
  • Contains an alpha-cyano group.
  • Targets sodium channels in neurons, causing delayed inactivation and membrane depolarization.
  • Manages a wide array of pests including fleas, cockroaches, flies, and ants.
  • Effective against malaria-transmitting Anopheles species.
  • Acts as a xenoestrogen, promoting human breast carcinoma cell proliferation *in vitro*.
  • Relevant to disease areas such as infection, neurological, and parasitic infection.
Catalog No. 50-004-05598 Supplier Medchemexpress LLC Supplier No. 250MGHYB0836
Add to Cart
Add to Cart
λ-Cyhalothrin is a highly efficient, broad-spectrum type II synthetic pyrethroid insecticide containing an α-cyano group. It is utilized to manage a wide array of pests across various applications. This neurotoxin targets sodium channels within the membranes of neurons in the central nervous system.
  • Highly efficient, broad-spectrum type II synthetic pyrethroid insecticide.
  • Contains an alpha-cyano group.
  • Targets sodium channels in neurons, causing delayed inactivation and membrane depolarization.
  • Manages a wide array of pests including fleas, cockroaches, flies, and ants.
  • Effective against malaria-transmitting Anopheles species.
  • Acts as a xenoestrogen, promoting human breast carcinoma cell proliferation *in vitro*.
  • Relevant to disease areas such as infection, neurological, and parasitic infection.