Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyhalothrin, ≥98.0% (mixture of 4 isomers, HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019771 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9771 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9771 Supplier MilliporeSigma Supelco Supplier No. 74767100MG
Add to Cart
Add to Cart

Specifications

CAS 68085-85-8
Linear Formula C23H19 ClF3 NO3
MDL Number MFCD00144031
Melting Point 48°C to 54°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 449.85
Percent Purity ≥98.0% (mixture of 4 isomers, HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.