Learn More
Strem, An Ascensus Company CAS# 247940-06-3. 5G. 2-(Dicyclohexylphosphino))-1,1'-biphenyl, 98% CyJohnPhos. MFCD01862441
SDP
Catalog No. 501963558
Encompass
CAS# 247940-06-3. 5G. 2-(Dicyclohexylphosphino))-1,1'-biphenyl, 98% CyJohnPhos. MFCD01862441. Molecular Formula: C24H31P. Molecular Weight: 350.49. Color/Form: white xtl. Strem# 15-1140. www.strem.com/catalog/v3/15-1140/
Catalog No. 50-196-3558 Supplier Strem, An Ascensus Company Supplier No. 1511405G
CAS# 247940-06-3. 5G. 2-(Dicyclohexylphosphino))-1,1'-biphenyl, 98% CyJohnPhos. MFCD01862441. Molecular Formula: C24H31P. Molecular Weight: 350.49. Color/Form: white xtl. Strem# 15-1140. www.strem.com/catalog/v3/15-1140/