Learn More
Enzo Life Sciences CYLD (human) polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW07600025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 336-353 of human CYLD. Source: Purified from rabbit serum. Species reactivity: Human. Applications: WB. Purity: Purified by salt precipitation. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1552