Learn More
Perkin Elmer US LLC NIR Quartz SUPRASIL 300 Macro Cylindrical Cell with PTFE Stopper - 10 mm, Qty. 1
Catalog No. 509042657
Encompass_Preferred
Cylindrical cells have an outside diameter of 22 mm and an inside diameter of 19 mm. A package contains one cell and two windows. NIR Quartz SUPRASIL 300 cells are made of synthetic quartz free from OH absorption. They have a spectral range of 200 - 3,500 nm and are identified by a red PerkinElmer logo.
Catalog No. 50-904-2657 Supplier Perkin Elmer US LLC Supplier No. B0631099
Cylindrical cells have an outside diameter of 22 mm and an inside diameter of 19 mm. A package contains one cell and two windows. NIR Quartz SUPRASIL 300 cells are made of synthetic quartz free from OH absorption. They have a spectral range of 200 - 3,500 nm and are identified by a red PerkinElmer logo.