Learn More
Metrohm USA CYLINDER 10ML COMPLETE
SDP
Supplier:  Metrohm USA 61574210
Encompass
Glass cylinder unit complete with cylinder, piston, cylinder base, valve disk and centering tube. For dosing units.
Catalog No. NC0093354