Learn More
Chemglass Life Sciences Chemglass 250 mL PYREX graduated cylinder, red stripe, reinforcement bead
SDP
Catalog No. NC2943925
Encompass_Preferred
250 mL graduated cylinder for routine laboratory liquid measurement. Made from PYREX glass, it is calibrated "to contain" with a single metric scale and features a red identification stripe and a reinforced bead to help reduce breakage.
  • 250 mL capacity.
  • Calibrated "to contain" with a single metric scale.
  • Red identification stripe for quick visual identification.
  • Reinforcement bead near the rim to reduce breakage.
  • Sold individually (pack quantity: 1).
Catalog No. NC2943925 Supplier Chemglass Life Sciences Supplier No. CG8254250
250 mL graduated cylinder for routine laboratory liquid measurement. Made from PYREX glass, it is calibrated "to contain" with a single metric scale and features a red identification stripe and a reinforced bead to help reduce breakage.
  • 250 mL capacity.
  • Calibrated "to contain" with a single metric scale.
  • Red identification stripe for quick visual identification.
  • Reinforcement bead near the rim to reduce breakage.
  • Sold individually (pack quantity: 1).