Learn More
Grainger Approved Vendor Carbide Bur, Cylinder, Double Cut, 3/16 in. x 5/8 in., 1 ea.
Catalog No. 502785264
Encompass_Preferred
This carbide cylinder bur features an end-cut design with double-cut flutes for efficient stock removal and a smooth surface finish. It is engineered for versatile use in demanding machining environments, offering durability when working with various high-strength metals.
  • Double-cut flute geometry for high stock removal rates on steel, stainless steel, cast iron, and high-temperature alloys
  • Cylindrical head shape with flat end-cut profile for targeted material removal
  • Constructed with YG7 carbide head and steel shank for long-lasting performance
  • Compatible with die grinders, rotary power tools, and straight grinders
  • Maximum operating speed of 30,000 RPM
Catalog No. 50-278-5264 Supplier Grainger Supplier No. 19D813
This carbide cylinder bur features an end-cut design with double-cut flutes for efficient stock removal and a smooth surface finish. It is engineered for versatile use in demanding machining environments, offering durability when working with various high-strength metals.
  • Double-cut flute geometry for high stock removal rates on steel, stainless steel, cast iron, and high-temperature alloys
  • Cylindrical head shape with flat end-cut profile for targeted material removal
  • Constructed with YG7 carbide head and steel shank for long-lasting performance
  • Compatible with die grinders, rotary power tools, and straight grinders
  • Maximum operating speed of 30,000 RPM