Learn More
Vee Gee Cylinder, Graduated , Class A, 250 mL
Catalog No. NC2386747
Encompass_Preferred
SIBATA Class A Graduated Cylinder with double-metric scale, graduated, to contain, and calibration temperature of 20°C. The graduated cylinder features hexagonal base for added stability* Made out of high-quality borosilicate glass* Double metric scale* Contains durable white enamel graduation markings* Wide range of sizes available (5 mL to 2000 mL)* Feature hexagonal bases for added stability* All sizes (except 2000 mL) come with a protective bumper guard* Calibration Temperature: 68°F (20°C)* Exceeds ASTM E-1272 Class A Standards * Graduation Interval: 2 mL* Graduation Range: 10.0 to 250.0 mL* Material: Borosilicate Glass* Scale Type(s): Double Metric* Tolerance: ±1.00 mL* Quantity: Pack of 4
Catalog No. NC2386747 Supplier Vee Gee Supplier No. 2351A250
SIBATA Class A Graduated Cylinder with double-metric scale, graduated, to contain, and calibration temperature of 20°C. The graduated cylinder features hexagonal base for added stability* Made out of high-quality borosilicate glass* Double metric scale* Contains durable white enamel graduation markings* Wide range of sizes available (5 mL to 2000 mL)* Feature hexagonal bases for added stability* All sizes (except 2000 mL) come with a protective bumper guard* Calibration Temperature: 68°F (20°C)* Exceeds ASTM E-1272 Class A Standards * Graduation Interval: 2 mL* Graduation Range: 10.0 to 250.0 mL* Material: Borosilicate Glass* Scale Type(s): Double Metric* Tolerance: ±1.00 mL* Quantity: Pack of 4