Learn More
Chemglass Life Sciences 250 mL Class A Pyrex Graduated Cylinder, Double Metric Scale
SDP
Catalog No. NC2548639
Encompass_Preferred
This 250 mL Class A graduated cylinder is made from Pyrex borosilicate glass and is calibrated "To Deliver" to ASTM E-1272 and ASTM E-542. It features a double metric scale with durable blue enamel markings and is supplied with shock-absorbing polyethylene bumper guards for added protection in routine laboratory use.
  • Class A accuracy, calibrated "To Deliver" to ASTM E-1272 and ASTM E-542.
  • 250 mL nominal capacity.
  • Double metric scale, numbered up and down, with blue enamel markings.
  • Pyrex borosilicate glass construction.
  • Supplied with shock-absorbing polyethylene bumper guards.
Catalog No. NC2548639 Supplier Chemglass Life Sciences Supplier No. CG8248250
This 250 mL Class A graduated cylinder is made from Pyrex borosilicate glass and is calibrated "To Deliver" to ASTM E-1272 and ASTM E-542. It features a double metric scale with durable blue enamel markings and is supplied with shock-absorbing polyethylene bumper guards for added protection in routine laboratory use.
  • Class A accuracy, calibrated "To Deliver" to ASTM E-1272 and ASTM E-542.
  • 250 mL nominal capacity.
  • Double metric scale, numbered up and down, with blue enamel markings.
  • Pyrex borosilicate glass construction.
  • Supplied with shock-absorbing polyethylene bumper guards.