Learn More
Chemglass Life Sciences Chemglass CG-8240-25 Cylinder, Class A, 25 mL, Graduated, Double Metric Scale
SDP
Catalog No. NC1963981
Encompass_Preferred
Chemglass Class A 25 mL graduated cylinder for accurate laboratory liquid measurement. Calibrated "to deliver" to ASTM tolerances, the cylinder features a double metric scale and includes a shock-absorbing bumper for added durability and stability.
  • Capacity: 25 mL.
  • Class: Class A.
  • Calibration: To deliver.
  • Scale type: Double metric scale, numbered up and down, in durable enamel.
  • Material: Borosilicate glass.
  • Supplied with: Shock-absorbing polyethylene bumper guards.
  • Packaging: Pack of 1.
  • Standards: Meets tolerances in ASTM E-1272 and ASTM E-542.
Catalog No. NC1963981 Supplier Chemglass Life Sciences Supplier No. CG824025
Chemglass Class A 25 mL graduated cylinder for accurate laboratory liquid measurement. Calibrated "to deliver" to ASTM tolerances, the cylinder features a double metric scale and includes a shock-absorbing bumper for added durability and stability.
  • Capacity: 25 mL.
  • Class: Class A.
  • Calibration: To deliver.
  • Scale type: Double metric scale, numbered up and down, in durable enamel.
  • Material: Borosilicate glass.
  • Supplied with: Shock-absorbing polyethylene bumper guards.
  • Packaging: Pack of 1.
  • Standards: Meets tolerances in ASTM E-1272 and ASTM E-542.