Learn More
Vee Gee Cylinder, Graduated , Class A, 5 mL
Catalog No. NC2386756
Encompass_Preferred
SIBATA Class A Graduated Cylinder with double-metric scale, graduated, to contain, and calibration temperature of 20°C. The graduated cylinder features hexagonal base for added stability* Made out of high-quality borosilicate glass* Double metric scale* Contains durable white enamel graduation markings* Wide range of sizes available (5 mL to 2000 mL)* Feature hexagonal bases for added stability* All sizes (except 2000 mL) come with a protective bumper guard* Calibration Temperature: 68°F (20°C)* Exceeds ASTM E-1272 Class A Standards * Graduation Interval: 0.1 mL* Graduation Range: 0.5 to 5.0 mL* Material: Borosilicate Glass* Scale Type(s): Double Metric* Tolerance: ±0.05 mL* Quantity: Pack of 4
Catalog No. NC2386756 Supplier Vee Gee Supplier No. 2351A5
SIBATA Class A Graduated Cylinder with double-metric scale, graduated, to contain, and calibration temperature of 20°C. The graduated cylinder features hexagonal base for added stability* Made out of high-quality borosilicate glass* Double metric scale* Contains durable white enamel graduation markings* Wide range of sizes available (5 mL to 2000 mL)* Feature hexagonal bases for added stability* All sizes (except 2000 mL) come with a protective bumper guard* Calibration Temperature: 68°F (20°C)* Exceeds ASTM E-1272 Class A Standards * Graduation Interval: 0.1 mL* Graduation Range: 0.5 to 5.0 mL* Material: Borosilicate Glass* Scale Type(s): Double Metric* Tolerance: ±0.05 mL* Quantity: Pack of 4