Learn More
Chemglass Life Sciences Chemglass Class A 500 mL Pyrex Graduated Cylinder, Double Metric Scale, with Bumper Guard
SDP
Catalog No. NC2548640
Encompass_Preferred
A 500 mL Class A PYREX graduated cylinder calibrated "to deliver" for precise laboratory volume measurement and dispensing. The cylinder features a double metric scale with durable enamel graduations and a shock-absorbing polyethylene bumper guard to protect the base.
  • Capacity: 500 mL.
  • Cylinder class: Class A (to deliver).
  • Material: Borosilicate glass.
  • Scale: Double metric scale, numbered up and down.
  • Graduations: Durable blue enamel.
  • Calibration: ASTM E-1272; ASTM E-542.
  • Bumper guard: Shock-absorbing polyethylene.
  • Pack quantity: 1 per pack.
Catalog No. NC2548640 Supplier Chemglass Life Sciences Supplier No. CG8248500
A 500 mL Class A PYREX graduated cylinder calibrated "to deliver" for precise laboratory volume measurement and dispensing. The cylinder features a double metric scale with durable enamel graduations and a shock-absorbing polyethylene bumper guard to protect the base.
  • Capacity: 500 mL.
  • Cylinder class: Class A (to deliver).
  • Material: Borosilicate glass.
  • Scale: Double metric scale, numbered up and down.
  • Graduations: Durable blue enamel.
  • Calibration: ASTM E-1272; ASTM E-542.
  • Bumper guard: Shock-absorbing polyethylene.
  • Pack quantity: 1 per pack.