Learn More
Chemglass Life Sciences Cylinder, Class B, 1,000 mL, Graduated, Single Metric Scale
SDP
Catalog No. NC1966737
Encompass_Preferred
Class B graduated cylinder with 1,000 mL capacity, single metric scale, and calibrated "to deliver." Meets ASTM E1272, Style 1, Class B, for tolerance and accuracy and is supplied with a polyethylene bumper guard for protection.
  • 1,000 mL capacity, calibrated to deliver.
  • Class B accuracy per ASTM E1272, Style 1.
  • Borosilicate glass construction for chemical resistance.
  • Polyethylene bumper guard for protection and stability.
  • Graduation interval 10 mL; tolerance ±6.0 mL; approximate OD x height 64 x 465 mm.
Catalog No. NC1966737 Supplier Chemglass Life Sciences Supplier No. CG82421L
Class B graduated cylinder with 1,000 mL capacity, single metric scale, and calibrated "to deliver." Meets ASTM E1272, Style 1, Class B, for tolerance and accuracy and is supplied with a polyethylene bumper guard for protection.
  • 1,000 mL capacity, calibrated to deliver.
  • Class B accuracy per ASTM E1272, Style 1.
  • Borosilicate glass construction for chemical resistance.
  • Polyethylene bumper guard for protection and stability.
  • Graduation interval 10 mL; tolerance ±6.0 mL; approximate OD x height 64 x 465 mm.