Learn More
Metrohm USA Cylinder element ETFE 50 mL
SDP
Supplier:  Metrohm USA 61566250
Encompass
For dosing units used for dosing alkali reagents or hydrofluoric acid (HF) with Dosinos. Less precision and reproducibility is to be expected in comparison with glass cylinders.
Catalog No. NC3678218