Learn More
Chemglass Life Sciences Cylinder, Graduated, 250 mL, Single Metric Scale, To Contain (250 mL), PYREX
SDP
Catalog No. NC2959215
Encompass_Preferred
250 mL graduated cylinder with single metric scale and "to contain" calibration, made from borosilicate glass for laboratory volumetric measurement and liquid handling. Supplied individually and suitable for routine measurement and transfer tasks.
  • 250 mL capacity with clear enamel graduations.
  • Single metric scale for straightforward volume reading.
  • Calibrated to contain for accurate volume holding prior to transfer.
  • Made from borosilicate glass for chemical resistance and thermal stability.
  • Supplied individually; bumper guards included on sizes 25 mL-2 L for added protection.
Catalog No. NC2959215 Supplier Chemglass Life Sciences Supplier No. CG8238250
250 mL graduated cylinder with single metric scale and "to contain" calibration, made from borosilicate glass for laboratory volumetric measurement and liquid handling. Supplied individually and suitable for routine measurement and transfer tasks.
  • 250 mL capacity with clear enamel graduations.
  • Single metric scale for straightforward volume reading.
  • Calibrated to contain for accurate volume holding prior to transfer.
  • Made from borosilicate glass for chemical resistance and thermal stability.
  • Supplied individually; bumper guards included on sizes 25 mL-2 L for added protection.