Learn More
Chemglass Life Sciences Cylinder, Graduated, 25 mL, Single Metric Scale (To Contain)
SDP
Catalog No. NC2959213
Encompass_Preferred
A 25 mL borosilicate glass graduated cylinder with a single metric scale, calibrated "to contain" for routine laboratory volumetric measurements. The cylinder features white enamel graduations and is supplied with a shock-absorbing bumper guard for improved stability and protection.
  • Capacity: 25 mL.
  • Calibration: to contain.
  • Single metric scale with white enamel graduations.
  • Borosilicate glass construction for chemical and thermal resistance.
  • Supplied with a shock-absorbing bumper guard for stability.
  • Packaged as a single unit for individual use.
  • Reusable and suitable for routine laboratory measurements.
Catalog No. NC2959213 Supplier Chemglass Life Sciences Supplier No. CG823825
A 25 mL borosilicate glass graduated cylinder with a single metric scale, calibrated "to contain" for routine laboratory volumetric measurements. The cylinder features white enamel graduations and is supplied with a shock-absorbing bumper guard for improved stability and protection.
  • Capacity: 25 mL.
  • Calibration: to contain.
  • Single metric scale with white enamel graduations.
  • Borosilicate glass construction for chemical and thermal resistance.
  • Supplied with a shock-absorbing bumper guard for stability.
  • Packaged as a single unit for individual use.
  • Reusable and suitable for routine laboratory measurements.