Learn More
Hach Company Cylinder, Graduated, Funnel-Top, 500 mL, Certified
Supplier:  Hach Company 2636249
Encompass
Certified Class A, this glass cylinder is calibrated "to deliver" according to ASTM E542 & E1272. Serialized and traceable to NIST standards (includes Certificate of Identification and Capacity). Single metric scale inwhite marking with funnel-top. Bumper guard included for all sizes ≥ 25 mL.
Catalog No. 50-199-6417