Learn More
Wilmad Labglass CYLINDER GRAD CLS A TD 1000ML
Catalog No. 50805552
Encompass
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 1000mL; Subdivision: 10mL; Tolerance: 3.00mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.
Catalog No. 50-805-552 Supplier Wilmad Labglass Supplier No. LG5520112
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 1000mL; Subdivision: 10mL; Tolerance: 3.00mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.