Learn More
Wilmad Labglass CYLINDER GRAD CLS A TD 250ML
Catalog No. 50803648
Encompass
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 250mL; Subdivision: 2mL; Tolerance: 1.00mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.
Catalog No. 50-803-648 Supplier Wilmad Labglass Supplier No. LG5520108
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 250mL; Subdivision: 2mL; Tolerance: 1.00mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.