Learn More
Wilmad Labglass CYLINDER GRAD CLS A TD 25ML
Catalog No. 50803647
Encompass
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 25mL; Subdivision: 0.2mL; Tolerance: 0.17mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.
Catalog No. 50-803-647 Supplier Wilmad Labglass Supplier No. LG5520102
Cylinder, Graduated, Class 'A', To Deliver. Capacity: 25mL; Subdivision: 0.2mL; Tolerance: 0.17mL. Calibrated to deliver at 20degreeC. ConForms to ASTM E 1272.