Learn More
Wilmad Labglass CYLINDER GRAD CLS B TD 1000ML
Catalog No. 50806372
Encompass_Preferred
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 1000mL; Subdivisions: 10mL; Tolerance:6.0mL, Approximate Height: 460mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.
Catalog No. 50-806-372 Supplier Wilmad Labglass Supplier No. LG5510114
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 1000mL; Subdivisions: 10mL; Tolerance:6.0mL, Approximate Height: 460mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.