Learn More
Wilmad Labglass CYLINDER GRAD CLS B TD 100ML
Catalog No. 50806211
Encompass_Preferred
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 100mL; Subdivisions: 1mL; Tolerance: 1.0mL, Approximate Height: 255mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.
Catalog No. 50-806-211 Supplier Wilmad Labglass Supplier No. LG5510108
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 100mL; Subdivisions: 1mL; Tolerance: 1.0mL, Approximate Height: 255mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.