Learn More
Wilmad Labglass CYLINDER GRAD CLS B TD 250ML
Catalog No. 50803482
Encompass_Preferred
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 250mL; Subdivisions: 2mL; Tolerance: 2.0mL, Approximate Height: 330mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.
Catalog No. 50-803-482 Supplier Wilmad Labglass Supplier No. LG5510110
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 250mL; Subdivisions: 2mL; Tolerance: 2.0mL, Approximate Height: 330mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.