Learn More
Wilmad Labglass CYLINDER GRAD CLS B TD 4000ML
Catalog No. 50805001
Encompass_Preferred
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 4000mL; Subdivisions: 50mL; Tolerance: 29.0mL, Approximate Height: 585mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.
Catalog No. 50-805-001 Supplier Wilmad Labglass Supplier No. LG5510118
Cylinder, Graduated, Laboratory Grade, To Deliver. Capacity: 4000mL; Subdivisions: 50mL; Tolerance: 29.0mL, Approximate Height: 585mm. Calibrated to deliver at 20degreeC and suitable For general laboratory use.