Learn More
Cole-Parmer CYLINDER GRAD HEX 100ML 1PK
Supplier:  Cole-Parmer 3450272
Encompass_Preferred
Graduated Cylinder, Glass, Hexagonal Base, 100 mL; 1/PK. Hexagonal base prevents cylinder from tipping over; Durable Borosilicate 3.3 Glass; Easy read graduations; Autoclavable
Catalog No. NC2394398