Learn More
Hach Company Cylinder, Graduated, Mixing, Glass, 25 mL +-0.3 mL, 0.5 mL divisions, glass stopper #13
Catalog No. 501995296 Shop All Hach Company Products
Encompass
This glass graduated cylinder is calibrated “to contain” with a molded in base. Each includes glass penny head stopper. Graduations are in durable white enamel bake.
Catalog No. 50-199-5296 Supplier Hach Company Supplier No. 2088640
This glass graduated cylinder is calibrated “to contain” with a molded in base. Each includes glass penny head stopper. Graduations are in durable white enamel bake.