Learn More
Hach Company Cylinder, Graduated, Mixing, 100 mL, Certified
Supplier:  Hach Company 2636342
Encompass
This certified, Class A glass mixing cylinder is calibrated “to contain” according to ASTM E542 & E1272. Serialized and traceable to NIST standards (includes Certificate of Identification and Capacity). Single metric scale in durable white markings with funnel-top. Glass penny head stopper included.
Catalog No. 50-199-6422