Learn More
Chemglass Life Sciences 250mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale
SDP
Catalog No. 501214719
Encompass_Preferred
250mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale. Cylinders are manufactured from polypropylene. They meet Class B specifications per DIN 12681. Cylinders have ring molded graduations and have blue screening to simplify the measurements.
Catalog No. 50-121-4719 Supplier Chemglass Life Sciences Supplier No. CLS1776250
250mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale. Cylinders are manufactured from polypropylene. They meet Class B specifications per DIN 12681. Cylinders have ring molded graduations and have blue screening to simplify the measurements.