Learn More
Bellco Glass, Inc. Cylinder Graduated Polypropylene,25mL
SDP
Catalog No. 501945734
Encompass
Cylinders are useful for general laboratory procedures and are autoclavable. They have ring molded graduationsfor increased durability. Material: polypropylene Autoclavable Size: 25ml Dimensions: 21x169mm
Catalog No. 50-194-5734 Supplier Bellco Glass, Inc. Supplier No. 562610025
Cylinders are useful for general laboratory procedures and are autoclavable. They have ring molded graduationsfor increased durability. Material: polypropylene Autoclavable Size: 25ml Dimensions: 21x169mm