Learn More
Chemglass Life Sciences 25mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale
SDP
Catalog No. 501214677
Encompass_Preferred
25mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale. Cylinders are manufactured from polypropylene. They meet Class B specifications per DIN 12681. Cylinders have ring molded graduations and have blue screening to simplify the measurements.
Catalog No. 50-121-4677 Supplier Chemglass Life Sciences Supplier No. CLS1776025
25mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale. Cylinders are manufactured from polypropylene. They meet Class B specifications per DIN 12681. Cylinders have ring molded graduations and have blue screening to simplify the measurements.