Learn More
Bellco Glass, Inc. Cylinder Graduated Polypropylene,500mL
SDP
Catalog No. 501945738
Encompass
Cylinders are useful for general laboratory procedures and are autoclavable. They have ring molded graduationsfor increased durability. Material: polypropylene Autoclavable Size: 500ml Dimensions: 61x350mm
Catalog No. 50-194-5738 Supplier Bellco Glass, Inc. Supplier No. 562610500
Cylinders are useful for general laboratory procedures and are autoclavable. They have ring molded graduationsfor increased durability. Material: polypropylene Autoclavable Size: 500ml Dimensions: 61x350mm