Learn More
Chemglass Life Sciences 500mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale
SDP
Catalog No. 501214746
Encompass_Preferred
500mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale. Cylinders are manufactured from polypropylene. They meet Class B specifications per DIN 12681. Cylinders have ring molded graduations and have blue screening to simplify the measurements.
Catalog No. 50-121-4746 Supplier Chemglass Life Sciences Supplier No. CLS1776500
500mL Cylinder, Graduated, Class B, Polypropylene, Blue Scale. Cylinders are manufactured from polypropylene. They meet Class B specifications per DIN 12681. Cylinders have ring molded graduations and have blue screening to simplify the measurements.