Learn More
Chemglass Life Sciences Cylinder, Graduated, 500mL, Single Scale, To Contain
SDP
Catalog No. NC1448467
Encompass_Preferred
Cylinder, Graduated, 500mL, Single Scale, To Contain. Calibrated “To Contain”, with white enamel graduations. The 100mL capacity is in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86, D-216 and D-447. The 10mL size has a funnel top. Bumper guards are supplied with 25mL thru 2L sizes inclusive.
Catalog No. NC1448467 Supplier Chemglass Life Sciences Supplier No. CG8238500
Cylinder, Graduated, 500mL, Single Scale, To Contain. Calibrated “To Contain”, with white enamel graduations. The 100mL capacity is in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86, D-216 and D-447. The 10mL size has a funnel top. Bumper guards are supplied with 25mL thru 2L sizes inclusive.