Learn More
Hach Company Cylinder, Graduated, 10 mL +- 0. 1 mL, 0.2 mL divisions (white markings)
Catalog No. 501994345 Shop All Hach Company Products
Encompass
Calibrated "to deliver" glass cylinder. Single metric scale in durable white markings.
Catalog No. 50-199-4345 Supplier Hach Company Supplier No. 50838
Calibrated "to deliver" glass cylinder. Single metric scale in durable white markings.