Learn More
Hach Company Cylinder, Graduated, 50 mL +/- 0.4 mL, 1.0 mL divisions (white markings)
Catalog No. 501994347 Shop All Hach Company Products
Encompass
Calibrated "to contain" glass cylinder. Single metric scale in durable white markings.
Catalog No. 50-199-4347 Supplier Hach Company Supplier No. 50841
Calibrated "to contain" glass cylinder. Single metric scale in durable white markings.