Learn More
Hach Company Cylinder, Graduated, Polycarbonate, 500 mL, 5.0 mL divisions
Catalog No. 501995441 Shop All Hach Company Products
Encompass
Clear polycarbonate cylinder is autoclavable and withstands temperatures to 138°C (280°F). Features easy to read graduations in durable fused ink.
Catalog No. 50-199-5441 Supplier Hach Company Supplier No. 2117949
Clear polycarbonate cylinder is autoclavable and withstands temperatures to 138°C (280°F). Features easy to read graduations in durable fused ink.