Learn More
Hach Company Cylinder, Graduated, Polycarbonate, 50 mL, 0.5 mL divisions
Catalog No. 501995438 Shop All Hach Company Products
Encompass
Clear polycarbonate cylinder is autoclavable and withstands temperatures to 138°C (280°F). Features easy to read graduations in durable fused ink.
Catalog No. 50-199-5438 Supplier Hach Company Supplier No. 2117941
Clear polycarbonate cylinder is autoclavable and withstands temperatures to 138°C (280°F). Features easy to read graduations in durable fused ink.