Learn More
Hach Company Cylinder, Graduated, Polymethylpentene, 100 mL +-0.6 mL, 1.0 mL divisions
Catalog No. 501994709 Shop All Hach Company Products
Encompass
This cylinder features transparent, nontoxic,noncontaminating, and biologically inert material with excellent chemical resistance. Meets ASTM E1272 Class B accuracy requirements, ISO Standard 6706, and suitable for food and beverage use (PMP resin meets requirements of CFR 21, Part 177.1520). Calibrated “To Contain/To Deliver” at 20°C (68°F).Shatterproof and thermally resistant to 135°C (275°F). Absence of meniscus for easy-to-read levels on front-to-back raised graduations.
Catalog No. 50-199-4709 Supplier Hach Company Supplier No. 217242
This cylinder features transparent, nontoxic,noncontaminating, and biologically inert material with excellent chemical resistance. Meets ASTM E1272 Class B accuracy requirements, ISO Standard 6706, and suitable for food and beverage use (PMP resin meets requirements of CFR 21, Part 177.1520). Calibrated “To Contain/To Deliver” at 20°C (68°F).Shatterproof and thermally resistant to 135°C (275°F). Absence of meniscus for easy-to-read levels on front-to-back raised graduations.