Learn More
Hach Company Cylinder, Graduated, Polymethylpentene, 250 mL +-1.4 mL, 2.0 mL divisions
Catalog No. 501994710 Shop All Hach Company Products
Encompass
This cylinder features transparent, nontoxic,noncontaminating, and biologically inert material with excellent chemical resistance. Meets ASTM E1272 Class B accuracy requirements, ISO Standard 6706, and suitable for food and beverage use (PMP resin meets requirements of CFR 21, Part 177.1520). Calibrated “To Contain/To Deliver” at 20°C (68°F).Shatterproof and thermally resistant to 135°C (275°F). Absence of meniscus for easy-to-read levels on front-to-back raised graduations.
Catalog No. 50-199-4710 Supplier Hach Company Supplier No. 217246
This cylinder features transparent, nontoxic,noncontaminating, and biologically inert material with excellent chemical resistance. Meets ASTM E1272 Class B accuracy requirements, ISO Standard 6706, and suitable for food and beverage use (PMP resin meets requirements of CFR 21, Part 177.1520). Calibrated “To Contain/To Deliver” at 20°C (68°F).Shatterproof and thermally resistant to 135°C (275°F). Absence of meniscus for easy-to-read levels on front-to-back raised graduations.