Learn More
Hach Company Cylinder, Graduated, Polypropylene, 250 mL +-1.4 mL, 2.0 mL divisions
Catalog No. 501994535 Shop All Hach Company Products
Encompass
Polypropylene construction of both cylinder and base. All sizes have generous pour spout, large molded-in, and easy-to-read graduations. Can be chemically sterilized without affecting accuracy. Autoclavable, but accuracy will be adversely affected. Cylinders meet accuracy requirements of ASTM Class B, E1272, ISO Standard 6706, and CFR 21, Part 177.1520
Catalog No. 50-199-4535 Supplier Hach Company Supplier No. 108146
Polypropylene construction of both cylinder and base. All sizes have generous pour spout, large molded-in, and easy-to-read graduations. Can be chemically sterilized without affecting accuracy. Autoclavable, but accuracy will be adversely affected. Cylinders meet accuracy requirements of ASTM Class B, E1272, ISO Standard 6706, and CFR 21, Part 177.1520