Learn More
Hach Company Cylinder, Graduated, 100 mL +-2.0 mL, 2.0mL divisions (low-form Tuttle, two spouts)
Catalog No. 501995210 Shop All Hach Company Products
Encompass
Tuttle-type glass cylinder calibrated "to deliver". Features two pour spouts and a single white metric scale.
Catalog No. 50-199-5210 Supplier Hach Company Supplier No. 2064342
Tuttle-type glass cylinder calibrated "to deliver". Features two pour spouts and a single white metric scale.