Learn More
Hach Company Cylinder, Graduated, 1000 mL, with Handles
Supplier:  Hach Company 2612953
Encompass
One-piece polypropylene construction with a molded in handle top ensure a secure, solid grip and superior control for safer, more convenient pouring. The “Total Contained, Total Delivered ”double scale is accurate and easy to read (cylinder will not create a meniscus), while the hexagonal base prevents tipping or rolling. Hand opening is 89x 29 mm (3 x 11/8"). Steam autoclavable upto 121°C (250°F).
Catalog No. 50-199-6349