Learn More
Hach Company Cylinder, Graduated, Double Metric, 1 L
Supplier:  Hach Company 2928305
Encompass
These glass graduated cylinders feature two sets of graduations for reading either up or down Calibrated "to deliver." Markings are white.
Catalog No. 50-199-6814